Luminaris color HiGreen qPCR master mix
Toimittaja: Thermo Fisher Scientific
K0361 K0362 K0363 K0364 K0371 K0372 K0373 K0374 K0391 K0392 K0393 K0394
K0361EA 299 EUR
K0361 K0362 K0363 K0364 K0371 K0372 K0373 K0374 K0391 K0392 K0393 K0394
Luminaris color HiGreen qPCR master mix
Nukleiinihapporeagenssit qPCR/RT-qPCR Enzymes and Kits
Luminaris Color HiGreen qPCR Master Mixes are universal ready to use solutions optimised for qPCR and two-step RT-qPCR. The master mixes include hot start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimised PCR buffer. Only the template and primers need to be added. The SYBR® Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.
  • UDG provides built-in carry-over contamination control
  • Colour change system to track pipetting
  • Versions with pre-mixed ROX or fluorescein for different real time platforms
The Hot Start Taq DNA polymerase in combination with an optimised buffer ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color HiGreen qPCR Master Mixes are supplemented with an inert blue dye and a separate yellow sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.
The use of Luminaris Color HiGreen qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color HiGreen qPCR Master Mixes with ROX at different concentrations or fluorescein as passive reference dyes are available for most qPCR platforms.