Luminaris color Probe qPCR master mixes
Toimittaja: Thermo Fisher Scientific
K0331 K0332 K0333 K0334 K0341 K0342 K0343 K0344 K0351 K0352 K0353 K0354
K0331EA 272 EUR
K0331 K0332 K0333 K0334 K0341 K0342 K0343 K0344 K0351 K0352 K0353 K0354
Luminaris color Probe qPCR master mixes
Nukleiinihapporeagenssit qPCR/RT-qPCR Enzymes and Kits
Luminaris Color Probe qPCR Master Mixes are universal ready-to-use solutions optimised for qPCR and two-step RT-qPCR. The master mixes include hot start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimised PCR buffer. Only the template, primers and probe need to be added.
  • UDG provides built-in carry-over contamination control
  • Colour change system to track pipetting
  • Versions with pre-mixed ROX for different real time platforms
The hot start Taq DNA polymerase, in combination with an optimised buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate yellow sample buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.
The use of Luminaris Color Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.