PCR master mix (2X)
Toimittaja: Thermo Fisher Scientific
K0171 K0172
K0171EA 132 EUR
K0171 K0172
PCR master mix (2X)
Nukleiinihapporeagenssit End-point PCR Enzymes and Kits
PCR Master Mix is a 2X concentrated solution of Taq DNA polymerase, dNTPs and all other components required for PCR, except DNA template and primers. This pre-mixed formulation saves time and reduces contamination due to a reduced number of pipetting steps required for PCR set up. The mix is optimised for efficient and reproducible PCR.