Biohazard Waste Containers
Toimittaja: JUSTRITE MANUFACTURING
05930WH 05930RD
JUSU05930WHEA 229 EUR
JUSU05930WH JUSU05930RD
Biohazard Waste Containers
Jäteastiat ja -säiliöt Jätesäiliöt
These leakproof sturdy steel containers with foot-operated and self-closing lid may be used to store contaminated laundry as well as other regulated waste (not for sharps). They have a biohazard bag which is held securely around the circumference of the can top.
Sertifikaatit: Comply with US OSHA Standard 29 CFR Part 1910.1030 for segregation of infectious waste. FM and TUV approved.