Dust filters, PressToCheck™
Toimittaja: JSP
BMN990-001-700 BMN750-000-600 BMN740-000-600
JSPABMN990-001-700EA 28.9 EUR
JSPABMN990-001-700 JSPABMN750-000-600 JSPABMN740-000-600
Dust filters, PressToCheck™
Suodattimet Ilmansuodattimet
PressToCheck™ twin filters for the Force®8 half mask and Force®10 full face mask facilitate instant and accurate checking of the face fit before entering the hazard area, without affecting the seal of the mask to the face.
  • Longer life/greater filter efficiency
  • Better protection in wet environments
  • Less chance of damage from foreign objects
  • Resistance to clogging from very fine dust environments
  • Improved hygiene
PressToCheck™ twin filters feature a bayonet fitting that makes the filters easy to attach. Lightweight, easy to wear, and available as dustMore Product Information