9000 Safety spectacle, Stealth™
Toimittaja: JSP
ASA770-15N-800 ASA770-15P-900 ASA770-16P-000 ASA770-16R-100 ASA770-15W-900 ASA770-16X-600
JSPAASA770-15N-800EA 20.1 EUR
JSPAASA770-15N-800 JSPAASA770-15P-900 JSPAASA770-16P-000 JSPAASA770-16R-100 JSPAASA770-15W-900 JSPAASA770-16X-600
9000 Safety spectacle, Stealth™
Suojalasit
The Stealth™ 9000 is a lightweight safety spectacle, weighing just 25 g, with a wraparound 9 base curve polycarbonate lens.
  • Lightweight safety spectacle weighing just 25 g
  • Adjustable nose bridge
  • UVA, UVB, and UVC protection
  • Class 1 optics with PremierShield™ (anti-scratch and anti-mist)
  • PVC nose pad
  • Polycarbonate frame
The Stealth™ 9000 safety spectacle features slim stylish temples that have been co-injection moulded with soft rubber to increase grip and provide excellent comfort levels. All of the Stealth™ 9000 safety spectacles come with class 1More Product Information