Visors and carriers for Evolution® safety helmets, Surefit™
Toimittaja: JSP
ANV000-001-108 ANX010-230-000 ANX060-230-000 ANW060-230-008
JSPAANV000-001-108EA 32.7 EUR
JSPAANV000-001-108 JSPAANX010-230-000 JSPAANX060-230-000 JSPAANW060-230-008
Visors and carriers for Evolution® safety helmets, Surefit™
Kypärät ja kolhulippikset Safety Helmets
Visor carrier clips into universal slot ear defender housing for the Evolution® safety helmet range. Available with polycarbonate or acetate visors to offer protection against high speed particles, liquid droplets and molten metal.
  • ABS visor carrier
  • Compatible with Evolution® helmet range
  • Can be fitted with polycarbonate or acetate visors
The 20 cm polycarbonate visor offers protection against medium energy, high speed particles at 120 m/s (270 mph), liquid droplets, and molten metal. The 20 cm acetate visor offers protection against low energy, highMore Product Information