Industrial visor range, EVOGuard®
Toimittaja: JSP
ADA240-060-000 ADA250-060-000 ADA260-070-000 ADA270-070-000 ADA280-070-000
JSPAADA240-060-000EA 37.3 EUR
JSPAADA240-060-000 JSPAADA250-060-000 JSPAADA260-070-000 JSPAADA270-070-000 JSPAADA280-070-000
Industrial visor range, EVOGuard®
Kasvosuojaimet
A range of high performance industrial visors, fully compatible with EVOGuard® browguard systems and EVO® helmet models.
  • Polycarbonate clear visor
  • Optical class 1 visor
  • Visors filter harmful UV radiation (UV 2C-1.2 protection)
  • Offering up to A-rated impact resistance
The optical class 1 visor offers a panoramic view with minimal distortion, ideal for comfortable all day wear. EVOGuard® visors maximise compatibility with JSP half masks, disposable respirators, prescription glasses, and the Sonis® helmet-mounted ear defender range. Visor filters harmfulMore Product Information