Frits, stainless steel, Unidense type
Toimittaja: VICI JOUR RESEARCH
JR-10FR2-5 JR-10FR1K-5 JR-10FR1-5 JR-10FR8-5 JR-.5FR8-5 JR-.5FR6-5 JR-.5FR4-5 JR-.5FR2-5 JR-.5FR1-5 JR-2FR2-5 JR-2FR8-5 JR-2FR4-5 JR-2FR1K-5 JR-2FR1-5 JR-1FR2-5 JR-1FR1-5
JOURJR-10FR2-5EA 0 EUR
JOURJR-10FR2-5 JOURJR-10FR1K-5 JOURJR-10FR1-5 JOURJR-10FR8-5 JOURJR-.5FR8-5 JOURJR-.5FR6-5 JOURJR-.5FR4-5 JOURJR-.5FR2-5 JOURJR-.5FR1-5 JOURJR-2FR2-5 JOURJR-2FR8-5 JOURJR-2FR4-5 JOURJR-2FR1K-5 JOURJR-2FR1-5 JOURJR-1FR2-5 JOURJR-1FR1-5
Frits, stainless steel, Unidense type
Suodattimet Esikolonnit ja fritit
Frits made of stainless steel with extra-low carbon content.
The sintering process used results in a strong bonding of the sinter metal particles. Depending upon the grade of powder used, the porosity of the frit can be precisely tuned.