Jenway® Cuvettes
Toimittaja: Cole-Parmer
83070-40 83070-34 99610-33 83070-76
JENW83070-40EA 204 EUR
JENW83070-40 JENW83070-34 JENW99610-33 JENW83070-76
Jenway® Cuvettes
Kyvetit Spektrofotometrin kyvetit
Get accurate spectroscopic readings.
  • Obtain consistent spectrophotometric data
Quartz and glass cuvettes are constructed of durable materials, making them ideal for repeated use. Single-use polystyrene cuvettes provide fast spectroscopic analysis and offer a less expensive alternative to glass and quartz cuvettes while eliminating washing and potential cross-contamination.