Kulta supplied in 0.1 mg/ml sodium citrate with stabilizer stabiloitu, nanoparticles 50 nm OD1 (Abs. 535 nm)

Toimittaja: Thermo Fisher Scientific

J67433.ACEA 218 EUR
J67433.AC J67433.AE
Kulta supplied in 0.1 mg/ml sodium citrate with stabilizer stabiloitu, nanoparticles 50 nm OD1 (Abs. 535 nm)
Kulta

CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pink to red MDL No.: MFCD00003436


Applications: For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Kaava: Au
Storage Temperature: Jääkaappi
MDL numero: MFCD00003436
CAS-numero: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR