Agarose, low gelling temperature

Toimittaja: Thermo Fisher Scientific
J66319.14 J66319.22 J66319.36
J66319.14EA 506 EUR
J66319.14 J66319.22 J66319.36
Agarose, low gelling temperature
Elektroforeesigeelit Agaroosi ja akryyliamidi
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

  • Powder form
  • Melting point 65 °C
  • Soluble in water
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR