Clomiphene citrate salt ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

J63663.04 J63663.09
J63663.04EA 163 EUR
J63663.04 J63663.09
Clomiphene citrate salt ≥98%
Clomiphene citrate salt

CAS No.: 50-41-9 Molecular Formula: C26H28ClNO.C6H8O7 Formula Weight: 598.08 Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00058322


Applications: A selective estrogen receptor modulator

Kaava: C₃₂H₃₆ClNO₈
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00058322
CAS-numero: 50-41-9
EINECS: 200-035-3
Merck Index: 14,2386

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR