Laemmli SDS sample buffer, non-reducing (4X)

Toimittaja: Thermo Fisher Scientific

J63615.ACEA 147 EUR
J63615.AC J63615.AD
Laemmli SDS sample buffer, non-reducing (4X)
Biokemialliset puskuriliuokset Elektroforeesipuskurit Loading Buffers

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR