Tween 20 washing buffer, 1% in PBS (10X)

Toimittaja: Thermo Fisher Scientific

J63314.AP J63314.K2 J63314.K3
J63314.APEA 156 EUR
J63314.AP J63314.K2 J63314.K3
Tween 20 washing buffer, 1% in PBS (10X)
Biokemialliset puskuriliuokset Elektroforeesipuskurit Washing Buffers

Storage Temperature: 2 to 8 °C
Physical Form: Viscous liquid

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR