Buffers, Sodium Citrate, 0,5M Solutions

Toimittaja: Thermo Fisher Scientific

J62918.AK J62918.AP J61815.AK J61815.AP J63888.AK J63888.AP J63199.AK J63199.AP
J62918.AKEA 168 EUR
J62918.AK J62918.AP J61815.AK J61815.AP J63888.AK J63888.AP J63199.AK J63199.AP
Buffers, Sodium Citrate, 0,5M Solutions
Biokemialliset puskuriliuokset Biological Buffers

Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00012462

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR