Myo-inositoli cell culture reagent

Toimittaja: Thermo Fisher Scientific

J62886.18 J62886.22 J62886.30
J62886.18EA 182 EUR
J62886.18 J62886.22 J62886.30
Myo-inositoli cell culture reagent

CAS No.: 87-89-8 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Crystalline powder Appearance: White Odour: Odourless Beilstein Reference: 1907329 Merck Reference: 14,4978 MDL No.: MFCD00077932


A lipotropic agent

Kaava: C₆H₁₂O₆
Kiehumispiste: 291 °C (1013 hPa)
Sulamispiste: 222…227 °C
Tiheys: 1,752 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00077932
CAS-numero: 87-89-8
EINECS: 201-781-2
Merck Index: 14,4978

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR