Magnesiumkloridi 1 M vesiliuos

Toimittaja: Thermo Fisher Scientific

J62411.ADEA 137 EUR
J62411.AD J62411.AE
Magnesiumkloridi 1 M vesiliuos
Magnesiumkloridi

CAS No.: 7786-30-3 Molecular Formula: Cl2Mg Formula Weight: 95.22 Physical Form: Liquid MDL No.: MFCD00011106

Kaava: MgCl₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011106
CAS-numero: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR