Etidiumbromidi 10 mg/ml aqueous solution

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 3,8-Diamino-5-etyyli-6-fenyylifenantridiumbromidi

J62282.AB
J62282.ABEA 166 EUR
J62282.AB
Etidiumbromidi 10 mg/ml aqueous solution
Etidiumbromidi
Kaava: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Kiehumispiste: 72
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011724
CAS-numero: 1239-45-8
EINECS: 214-984-6
UN: 2810
ADR: 6.1 III
Merck Index: 14,4731

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR