Orange G/blue loading dyes (6X)
Toimittaja: Thermo Fisher Scientific
J61877.AB J61877.AC
J61877.ABEA 187 EUR
J61877.AB J61877.AC
Orange G/blue loading dyes (6X)
Värit Elektroforeesivärit
Orange G/blue loading dye (6X) is used for loading DNA samples into gel electrophoresis wells and tracking migration during electrophoresis.
Used in molecular biology protocols, in nucleic acid gel electrophoresis and blotting to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.
Tilaustiedot: Contains 0,25% Orange G, 0,25% bromophenol blue, 0,25% xylene cyanol FF, 15% Ficoll®, 30 mM EDTA in autoclaved DI water.