Agaroosi, high melting , medium resolution for DNA separations

Toimittaja: Thermo Fisher Scientific

J61123.14 J61123.22 J61123.36
J61123.14EA 145 EUR
J61123.14 J61123.22 J61123.36
Agaroosi, high melting , medium resolution for DNA separations
Agaroosi

CAS No.: 9012-36-6 Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00081294


Applications: For standard DNA applications

Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081294
CAS-numero: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR