Histamine diphosphate 98%

Toimittaja: Thermo Fisher Scientific
Vaara

J61088.06 J61088.14
J61088.06EA 164 EUR
J61088.06 J61088.14
Histamine diphosphate 98%
Histamine diphosphate

Purity: 99+% CAS No.: 51-74-1 Molecular Formula: C5H9N3.2H3PO4 Formula Weight: 307.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 128-132° MDL No.: MFCD00064316


Applications: Endogenous H1 and H2 agonist

Kaava: C₅H₉N₃·2H₃PO₄
Sulamispiste: 128…132 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00064316
CAS-numero: 51-74-1
EINECS: 200-118-4
Merck Index: 14,4719

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR