Phosphatase inhibitor cocktail II

Toimittaja: Thermo Fisher Scientific

J61022.AA
J61022.AAEA 335 EUR
J61022.AA
Phosphatase inhibitor cocktail II
Soluviljelyyn suplementit ja lisäaineet

The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts.


  • Storage temperature: –30 to –10 °C
  • Physical form: Liquid
  • Appearance: Colourless


This product has been optimized and tested for tyrosine protein phosphatases, acid and alkaline phosphatases. It has been tested on cell extracts from animal tissues including bovine liver and human placenta, and cytosolic extract of rabbit muscle. It is a featured product for Kinase Phosphatase Biology research.An inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases. It is used with tissue and cell extracts, including extracts containing detergents.


Toimitustiedot: Contains 100 mM sodium fluoride, 50 mM sodium orthovanadate, 50 mM sodium pyrophosphate, and 100 mM b-glycerophosphate. 100X concentrate.


Varoitus: This cocktail will inhibit broad spectrum of serine/theorine and tyrosine phosphatases and is suitable for cell and tissue lysis. Store at –20 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR