Kanamysiinisulfaatti cell culture reagent

Toimittaja: Thermo Fisher Scientific
Vaara

J60668.03 J60668.06
J60668.03EA 131 EUR
J60668.03 J60668.06
Kanamysiinisulfaatti cell culture reagent
Kanamysiinisulfaatti

CAS No.: 25389-94-0 Molecular Formula: C18H36N4O11.H2SO4 Formula Weight: 582.58 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in water at 10mg/ml MDL No.: MFCD00070253

Kaava: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00070253
CAS-numero: 25389-94-0
EINECS: 246-933-9
Merck Index: 14,5281

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR