TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98% molekyylibiologiaan

Toimittaja: Thermo Fisher Scientific
Vaara

J60316.06 J60316.09 J60316.3S
J60316.06EA 406 EUR
J60316.06 J60316.09 J60316.3S
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98% molekyylibiologiaan
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Applications: For reducing disulfide bonds in peptides and proteins.


Applications: For reducing disulfide bonds in peptides and proteins.

Kaava: C₉H₁₆ClO₆P
Sulamispiste: 177…178 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00145469
CAS-numero: 51805-45-9
UN: 3261
ADR: 8,II
Merck Index: 14,9085

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR