Loratadine ≥98%

Toimittaja: Thermo Fisher Scientific

J60190.ME J60190.03 J60190.06
J60190.MEEA 201 EUR
J60190.ME J60190.03 J60190.06
Loratadine ≥98%
Loratadine

Purity: 98+% CAS No.: 79794-75-5 Molecular Formula: C22H23ClN2O2 Formula Weight: 382.88 Physical Form: Powder Appearance: White Melting Point: 134° MDL No.: MFCD00672869


Applications: A nonsedating-type histamine H1-receptor antagonist

Kaava: C₂₂H₂₃ClN₂O₂
Sulamispiste: 134 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00672869
CAS-numero: 79794-75-5
Merck Index: 14,5578

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR