Buffers, citrate, 0,5 M solutions

Toimittaja: Thermo Fisher Scientific

J60754.AP J60754.AK J60024.AK J60024.AP J60125.AK J60125.AP J61690.AK J61690.AP J63008.AK J63008.AP J61249.AK J61249.AP J61391.AK J61391.AP J63950.AP J63950.AK
J60754.APEA 254 EUR
J60754.AP J60754.AK J60024.AK J60024.AP J60125.AK J60125.AP J61690.AK J61690.AP J63008.AK J63008.AP J61249.AK J61249.AP J61391.AK J61391.AP J63950.AP J63950.AK
Buffers, citrate, 0,5 M solutions
Biokemialliset puskuriliuokset Biological Buffers

CAS No.: 6132-04-3
Storage temperature: 2 to 8 °C
Appearance: colourless
Odour: odourless
MDL No.: MFCD00150031

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR