Vista Sterile Cleanroom Overboots
Toimittaja: ISOFIELD IRELAND
80761
ISOF80761EA 413 EUR
ISOF80761
Vista Sterile Cleanroom Overboots
Kertakäyttöiset vaatteet
The Isofield Vista sterile cleanroom overboots are 50 cm (19,5”) tall and aseptically folded.
  • Sterile, cleanroom overboots
  • Extra-long design, 50 cm knee height
  • Two ties for added security
  • Anti-slip PU sole
  • Aseptically folded
Providing essential protection for GMP Grade A/B cleanroom applications, these anti-static, low-linting overboots are constructed with a non-slip PU sole, two sets of ties for secure fit, and excellent barrier properties.
Pakkaus:
Overboots are packaged 1 pair per inner bag, 10 pairs per outer bag, 8 outer bags per carton, 80 pairs per carton.