Gravimetric diluter, DiluFlow® Elite
Toimittaja: INTERSCIENCE
503101 503105 503201 503205
INSC503101EA 0 EUR
INSC503101 INSC503105 INSC503201 INSC503205
Gravimetric diluter, DiluFlow® Elite
Laimentimet Gravimetriset laimentimet
DiluFlow® Elite automatically dilutes in seconds a solid sample with the appropriate weight of diluent with full traceability. Programmable and connectable with up to 6 diluents.
  • Ultra low profile: bag opening height is only 30 cm which makes it ideal for work in a laminar flow cabinet
  • 32 editable programs, bidirectional connectivity
  • Up to 6 pumps with 4 connectable pumps (Watson Marlow®)
  • GeckoGrip System: Holds the bag in all conditions
  • Light Code: Intuitive indicator of the status
  • Removable DripTray for easy cleaning, in case of spillage
  • Multi-dispensing