tempmate® S1 Single Use Temperature Data Logger, 15 min, L1 8 °C, H1 25 °C, SD 60 min
Toimittaja: tempmate
S1C15A79100
IMECS1C15A79100EA 461 EUR
IMECS1C15A79100
tempmate® S1 Single Use Temperature Data Logger, 15 min, L1 8 °C, H1 25 °C, SD 60 min
Dataloggerit
Pre-programmed. Outstanding single-use temperature logger for reliable cold chain monitoring. Integrated USB port and PDF and CSV generating software on-board.
  • Pre-programmed
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S1 loggers are specially designed for temperature monitoring in any cold chain logistics. As a pre-More Product Information