External centering rings, type KF
Toimittaja: Welch by Gardner Denver
701151 701143 701141
ILMV701151EA 11.2 EUR
ILMV701151 ILMV701143 ILMV701141
External centering rings, type KF
Sulkijakalvot ja -matot Tiivisterenkaat
External centering rings for double sided mechanism made of aluminium, with perbunan O-ring.