Kalorimetri, C 7000
Toimittaja: IKA
8800900 8801400 8800900 8801400
622-0018EA 48510 EUR
622-0018 IKAA8801400 IKAC8800900 IKAC8801400
Kalorimetri, C 7000
Kalorimetrit
The C 7000 calorimeter is designed to measure the calorific power of samples in a dry procedure.
  • High sampling frequency
  • Exact determination of the calorific power acc. to ISO 1928
  • Automatic identification of calorimetric bombs
  • C 7010 standard calorimetric bomb can be mounted on a C 14 disposable crucible
  • Connection interface for balance, printer and PC
The temperature is measured in the calorimetric bomb. Measurement periods are reduced between 3 and 7 minutes depending on the type of sample. With its special coding system, the calorimeter can manageMore Product Information