Baths and circulators, plastic baths
Toimittaja: IKA
8035000 8035300 8035400 8034900
IKAA8035000EA 2700 EUR
IKAA8035000 IKAA8035300 IKAA8035400 IKAA8034900
Baths and circulators, plastic baths
Kiertotermostaatit Avoimet termostaattihaudejärjestelmät Hauteet, joissa irrotettavat termostaattihauteet
The ICC compact immersion circulators allow flexible coupling to water baths of different volumes. They are designed for use with liquids up to temperatures of 150 °C. The convenient carrying handle and the compact design allow easy transportation as well as comfortable use. An economic and attractive solution for basic applications such as sample tempering and heating glassware.
  • Adjustable safety circuit
  • Switch from internal to external temperature with one key (only for control unit)
  • Optical and acoustic warning, example: for over heating
  • Powerful pressure/suction pump