Refrigerated and heating circulators, CBC series
Toimittaja: IKA
4167000 4165000
IKAA4167000EA 8750 EUR
IKAA4167000 IKAA4165000
Refrigerated and heating circulators, CBC series
Kiertotermostaatit Avoimet termostaattihaudejärjestelmät Lämmittävät/jäähdyttävät termostaattihauteet
The CBC 5 is a powerful refrigerated and heating circulator with strong heating and cooling power with an operating temperature range of −25 to +200 °C.
  • Industry-standard LED/TFT display for maximum visibility
  • Adjustable safety circuit
  • Visible and audible alarm
  • Safety drain valve
  • USB/RS232 interfaces (labworldsoft® which enables online updates of device software)
The units proven pressure and suction pump makes the circulator ideal for external applications. The high-quality insulation allows fast heat-up times and reduces heat input at lowMore Product Information