Reactor systems, EasySyn
Toimittaja: IKA
20113436 20113433 20113434 20113435 20113438 20113440 20113442 20113443
IKAA20113436EA 15160 EUR
IKAA20113436 IKAA20113433 IKAA20113434 IKAA20113435 IKAA20113438 IKAA20113440 IKAA20113442 IKAA20113443
Reactor systems, EasySyn
Laboratorion reaktorijärjestelmät
EasySyn is the reactor system for organic or aqueous syntheses, and the only reactor system for which you can get all the complementary components from a single source: Overhead stirrers, circulators, vacuum pumps and controllers, sensors and automation software. All from IKA and all perfectly matched to each other. With its double-walled reactor vessels from 500 to 5000 ml, EasySyn is a medium-sized and very flexible upscaling system. It ensures safe and reproducible chemical reactions.
  • Ideal for organic or aqueous syntheses
The EasySyn reactorMore Product Information