UTTD Control Life Science Solution Homogenising System
Toimittaja: IKA
10013296
IKAA10013296EA 1870 EUR
IKAA10013296
UTTD Control Life Science Solution Homogenising System
Homogenisaattorit Homogenisaattorit, pöytämalli
The UTTD control life science solution package was specially put together to meet the critical challenges of homogenising tissue, cartilage and bone samples: homogeneous comminution of the samples and protection against cross-contamination.
  • Single-use homogenisation of tissue samples without risk of cross-contamination
The package includes the powerful ULTRA-TURRAX® Tube Drive P control homogenisation system, which has been designed for universal use. In combination with the patented DT-20 disposable dispersing chambers with 20 ml volume and rotor-More Product Information