Closed cap, Dino-Lite
Toimittaja: Dino-Lite / IDCP B.V.
ZT-Z-CC1
IDCPZT-Z-CC1EA 10.2 EUR
IDCPZT-Z-CC1
Closed cap, Dino-Lite
Mikroskoopit
Closed cap addition for the polarising AM series (ZT models). Designed to help protect the lens from dust and more.
This snap-on cover fits the AM413ZT, AM4013MZT and AM7013MZT model. Not designed for use with the AD series or AM models without polarisation.