Handheld digital microscopes, Dino-Lite High Speed (USB 3.0)
Toimittaja: Dino-Lite / IDCP B.V.
AM73115MZT AM73115MZTL AM73915MZT AM73915MZTL AM73515MZT AM73515MZTL AM73115MTF
IDCPAM73115MZTEA 1410 EUR
IDCPAM73115MZT IDCPAM73115MZTL IDCPAM73915MZT IDCPAM73915MZTL IDCPAM73515MZT IDCPAM73515MZTL IDCPAM73115MTF
Handheld digital microscopes, Dino-Lite High Speed (USB 3.0)
Mikroskoopit
Dino-Lite models with USB 3.0 offer an image transfer speed of up to 45 fps at a 1280×960 resolution. USB 3.0 adds a new transfer mode called "SuperSpeed" (SS) capable of transferring data up to 5 Gbits/s (625 MB/s). SuperSpeed allows users to take advantage of the full power of the Dino-Lite Edge Series optics including improved colour accuracy and image quality.
  • Super speed USB 3.0 connection
  • Metal housing: Microscopes are ESD-safe
  • Edge series: High-quality optics provide a very sharp, bright and natural colour image with very low aberration and