Agaroosi standard, white powder for molecular biology

Toimittaja: MP Biomedicals

11AGAH0500 11AGAH0250 11AGAH0100
ICNA11AGAH0500EA 537 EUR
ICNA11AGAH0500 ICNA11AGAH0250 ICNA11AGAH0100
Agaroosi standard, white powder for molecular biology
Agaroosi

Standard agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
Standard agarose is a very low electroendosmosis agarose (EEO) which is recommended for sharp resolution of nucleic acid fragments greater than 400 bp and up to 8000 bp. This Standard Agarose may be used at concentrations between 0.8 % and 2 %. Southern and Northern transfers can be performed efficiently from them.


  • "Molecular Biology Grade
  • Suitable for both analytical and preparative electrophoresis procedures
  • Presentation: White Powder
  • DNAase/RNase Activity and inhibition of ligation and restriction enzymes not detected
  • Soluble in water (1.5 g plus 100 mL)
  • Storage at room temperature"

Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081294
CAS-numero: 9012-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity Test Passes
Gelling Temperature (1.5% gel) 36 ± 2°C
Melting Temperature (1.5% gel) ~89°C
Gel Strength (1% gel) ≥1200g/cm2
Electroendosmosis ≤0.12
Sulphate ≤0.14%
DNAase/RNase Activity None Detected
DNA Blotting Complete Transfer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR