Nucleic acid extraction, MPure™ Viral nucleic acids extraction kits, MP Biomedicals

Toimittaja: MP Biomedicals

117022170 117022300
ICNA117022170EA 346 EUR
ICNA117022170 ICNA117022300
Nucleic acid extraction, MPure™ Viral nucleic acids extraction kits, MP Biomedicals
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents

MPure™ Viral Nucleic Acids Extraction Kit is used with the MPure-12 instrument for extraction of Viral DNA or RNA from human biological specimens such as serum, plasma, and other cell-free fluids.

Order Now

TEKNISET TIEDOT

Kuvaus MPure™ Viral Nucleic Acid Extraction Kit 800 MPure™ Viral Nucleic Acid Extraction Kit
Environmentally Preferable
Hakemus
Target molecule Copurification of DNA & RNA,DNA,RNA Copurification of DNA & RNA,DNA,RNA
Refine target molecule Viral RNA,Viral DNA Viral RNA,Viral DNA
Sample type Biofluids (serum, plasma, saliva) Swabs,Biofluids (serum, plasma, saliva)
Method / Format Magnetic beads Magnetic beads
Prep size Mini Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR