Lysing kits for Fastprep®-24 5G, Lysing Matrix D, MP Biomedicals
Toimittaja: MP Biomedicals
116933025 116913100 116913500 116933050 116540434 116913050 116973100 116953010 116933005 116953050 116973025 116973050 116983001 116983010
ICNA116933025EA 264 EUR
ICNA116933025 ICNA116913100 ICNA116913500 ICNA116933050 ICNA116540434 ICNA116913050 ICNA116973100 ICNA116953010 ICNA116933005 ICNA116953050 ICNA116973025 ICNA116973050 ICNA116983001 ICNA116983010
Lysing kits for Fastprep®-24 5G, Lysing Matrix D, MP Biomedicals
Homogenisaattorit Homogenisaattorit, pöytämalli
Lysing Matrix D is a critical component of FastPrep® sample preparation system. Each impact-resistant 2 ml tube contains 1,4 mm ceramic spheres. Lysing Matrix D tubes have green caps and are found in the FastDNA™ Spin Kit for Plant and Animal Tissues and FastRNA™ Pro Green Kit for isolation of nucleic acids from plants and animals.