Lysing kits for Fastprep®-24 5G, Lysing Matrix B, MP Biomedicals
Toimittaja: MP Biomedicals
116911050 116951050 116951010 116931050 116931025 116911500 116981001 116540428 116540429 116971025 116981010
ICNA116911050EA 232 EUR
ICNA116911050 ICNA116951050 ICNA116951010 ICNA116931050 ICNA116931025 ICNA116911500 ICNA116981001 ICNA116540428 ICNA116540429 ICNA116971025 ICNA116981010
Lysing kits for Fastprep®-24 5G, Lysing Matrix B, MP Biomedicals
Homogenisaattorit Homogenisaattorit, pöytämalli
Lysing Matrix B is a critical component of FastPrep® sample preparation system. Each impact-resistant 2 ml tube contains 0,1 mm silica spheres. Lysing Matrix B tubes have blue caps and are found in the FastRNA™ Pro Blue Kit and FastProtein™ Blue Matrix. The fast and easy to use protocol ensures successful lysis with even the most challenging samples for high efficiency and reliability.