Grinding matrix for lysing kits, MP Biomedicals
Toimittaja: MP Biomedicals
116540424 116040801 116540034 116540412 116540422
ICNA116540424EA 126 EUR
ICNA116540424 ICNA116040801 ICNA116540034 ICNA116540412 ICNA116540422
Grinding matrix for lysing kits, MP Biomedicals
Helmet ja partikkelit Ceramic Beads
Ceramic sphere is a Zirconium oxide coated grinding matrix which is chemically inert and will not bind nucleic acids. These are components of lysing matrices.