Cleaning solution, ES 7X™, MP Biomedicals
Toimittaja: MP Biomedicals
097667193 097667194 097667195 097667198
ICNA097667193EA 75.3 EUR
ICNA097667193 ICNA097667194 ICNA097667195 ICNA097667198
Cleaning solution, ES 7X™, MP Biomedicals
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
Safe and effective cleaning solution for laboratory instruments and plastic, glassware.
  • Yellowish cloudy liquid completely soluble in water at any concentration
  • Drains from glassware in seconds
  • Contains powerful sequestering reagents
  • Ideal for cell cultureware and general labware
The 7X™ laboratory detergent is ideal for thorough cleaning of cultureware and general lab equipment including test tubes, petri dishes, culture flasks and cylinders, burettes, pipettes, slide covers, and hypodermic syringes. It shows a broad range of application other thanMore Product Information