Agarose, immunodiffusion grade, MP Biomedicals
Toimittaja: MP Biomedicals
08952012 08952014
ICNA08952012EA 1110 EUR
ICNA08952012 ICNA08952014
Agarose, immunodiffusion grade, MP Biomedicals
Elektroforeesigeelit Agaroosi ja akryyliamidi
Agarose is the most popular medium for immunoelectrophoresis because of the large pore size for rapid diffusion, and for low background staining by Coomassie Blue G stain. The low EEO and high gel strength is specifically selected and tested for immunoelectrophoresis and immunodiffusion.