D(+)-sakkaroosi ≥99.9%, Ultrapure

Toimittaja: MP Biomedicals

04821713 04821721
ICNA04821713EA 130 EUR
ICNA04821713 ICNA04821721
D(+)-sakkaroosi ≥99.9%, Ultrapure
D(+)-sakkaroosi

Heavy metals (as Pb): <5 ppm
Glucose: <0.1%
Arsenic: <0.001 ppm
DNAse and RNAse-free.

Kaava: C₁₂H₂₂O₁₁
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006626
CAS-numero: 57-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR