Acrylamide, electrophoresis grade, MP Biomedicals

Toimittaja: MP Biomedicals
04814340 04814349
ICNA04814340EA 58.1 EUR
ICNA04814340 ICNA04814349
Acrylamide, electrophoresis grade, MP Biomedicals
Elektroforeesigeelit Agaroosi ja akryyliamidi
Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers. It polymerizes in the presence of free radicals in aqueous solutions. It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.

  • Suitable for use in electrophoresis
  • Purity >99%
  • pH 5,0 to 7,0 in 1% aqueous solution
  • Can be stored at room tempetarure as a dessicate
  • Soluble in water (1% aqueous solution), methanol (155 g/100 ml), ethanol (86 g/100 ml), alcohol, acetone (63,1 g/100 ml); only minimally soluble in benzene or heptane
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR