Cesiumkloridi ≥99.999%, white powder, Ultrapure

Toimittaja: MP Biomedicals

04813061 04813063 04813069
ICNA04813061EA 166 EUR
ICNA04813061 ICNA04813063 ICNA04813069
Cesiumkloridi ≥99.999%, white powder, Ultrapure
Cesiumkloridi

Cesium chloride , an inorganic compound, is an important source of cesium ions in a variety of applications.


Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins. It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation.


Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA. Procedures have been reported for the purification of closed circular DNA by equilibrium centrifugation in both continuous and discontinuous CsCl-ethidium bromide gradients. Subsequently, the CsCl can be removed from the DNA preparation by either dialysis or precipitation.


Freely soluble in water (500 mg/ml), (50% aqueous-clear, colourless solution); soluble in ethanol, alcohol.

Kaava: CsCl
Sulamispiste: 646 °C
MDL numero: MFCD00010955
CAS-numero: 7647-17-8

Order Now

Testitulosten erittely

Purity 99.999%
OD260 nm (50% aq soln) <0.02
pH 5.5 - 6.5 (1% aq soln)
Barium ≤1 ppm
Magnesium ≤ 0.5 ppm
Potassium ≤2 ppm
Iron ≤ 1 ppm
Calcium ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR