Glysiini ≥99.5%

Toimittaja: MP Biomedicals

04808822 04808831 04808873
ICNA04808822EA 136 EUR
ICNA04808822 ICNA04808831 ICNA04808873
Glysiini ≥99.5%
Glysiini

Electrophoresis Grade Heavy metals (as Pb): 20 ppm Used for preparation of Tris-glycine buffers.

Kaava: NH₂CH₂COOH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008131
CAS-numero: 56-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR