Natriumnitriitti 97.0-101.0%, pale yellow powder USP

Toimittaja: MP Biomedicals
Vaara

0296030190 0296030191
ICNA0296030190EA 72.9 EUR
ICNA0296030190 ICNA0296030191
Natriumnitriitti 97.0-101.0%, pale yellow powder USP
Natriumnitriitti

Sodium nitrite is used in many industrial processes, in meat curing, coloring, and preserving, and as a reagent in analytical chemistry.


It is used therapeutically as an antidote in cyanide poisoning. The compound is toxic and mutagenic and will react in vivo with secondary or tertiary amines thereby producing highly carcinogenic nitrosamines.


Soluble in water (820 mg/mL at 20 °C), methanol (4.5 mg/mL), ethanol (3 mg/mL), ether (slightly), and NH3 (very soluble)

Kaava: NaNO₂
MDL numero: MFCD00011118
CAS-numero: 7632-00-0
UN: 1500
ADR: 5.1,III

Order Now

Testitulosten erittely

Identity Test Passes
Loss on Drying (4 hours over silica) ≤0.25%
Heavy Metals ≤0,002%
Assay 97,0 - 101,0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR